missing translation for 'onlineSavingsMsg'
Learn More

Tungsteno, solución estándar plasmática, Specpure™, W 1000 μg/ml, Thermo Scientific Chemicals

Catalog Number p-7082672
Click to view available options
Cantidad:
50 mL
100 mL
500 mL
Este artículo no se puede devolver. Vea la política de devoluciones
(NH4)2WO4 in H2O
MFCD00011461
Este artículo no se puede devolver. Vea la política de devoluciones
(NH4)2WO4 in H2O
MFCD00011461

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Tungsten, plasma standard solution, Specpure™
Concentración W 1000μg/ml
Cantidad 50 mL
Forma física Líquida

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'