missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH-Receptor, A-Chain Mouse, Clone: TSHRA/1402, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 7253-MSM2-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TSH-Receptor, A-Chain | |
Cytoplasmic | |
TSHRA/1402 | |
Unconjugated | |
XP_005268094 | |
Mouse | |
62kDa | |
Protein A/G Purified | |
RUO | |
7253 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
TSHR | |
Recombinant human TSHR, A-Chain protein | |
293T Cells. Thyroid gland | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |