missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20003-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The antibody recognizes the C-term of TRPS1.
This gene encodes a transcription factor that represses GATA-regulated genes and binds to a dynein light chain protein. Binding of the encoded protein to the dynein light chain protein affects binding to GATA consensus sequences and suppresses its transcriptional activity. Defects in this gene are a cause of tricho-rhino-phalangeal syndrome (TRPS) types I-III.Especificaciones
TRPS1 | |
Polyclonal | |
Unconjugated | |
TRPS1 | |
GC79, LGCR, TRPS1, Zinc finger protein GC79 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
299897, 7227, 83925 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q925H1, Q9UHF7 | |
TRPS1 | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |