Learn More
Brand™ Transferpette™ S Fixed Volume Pipette
Provides all features required for pipetting in life science applications: lightweight, rugged, precise and reliable. Brand™ Transferpette™ S Fixed Volume Pipettes are the perfect manual choice for demanding laboratory applications.
Marca: Brand™ 704764
Detalles adicionales : Peso : 0.19040kg
Descripción
- Effortless one-handed operation
- Clearly visible volume display
- Completely autoclavable at 121°e;C
- Calibration adjustment without tools
- Corrosion-resistant pistons
- UV-resistant
- Ergonomic finger rest for relaxed pipetting
- Supplied with DE-M marking, performance certificate, shelf/rack mount and silicone grease
Especificaciones
≤ 0.8%, ± 16µL | |
F-2000 | |
Red | |
Single channel manual pipette | |
1/Pk. |
DE-M | |
Autoclavable | |
1 | |
2000 μL |