missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trace Amine Receptor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-38762
Detalles adicionales : Peso : 0.00970kg
Descripción
Trace Amine Receptor 1 Polyclonal specifically detects Trace Amine Receptor 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Especificaciones
Trace Amine Receptor 1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q96RJ0 | |
TAAR1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: KEQARLISDANQKLQIGLEMKNGISQSKER | |
0.1 mL | |
GPCR | |
134864 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |