missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trace Amine Receptor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-88452
Detalles adicionales : Peso : 0.00970kg
Descripción
Trace Amine Receptor 1 Polyclonal specifically detects Trace Amine Receptor 1 in Human samples. It is validated for Western Blot.Especificaciones
Trace Amine Receptor 1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of human Trace Amine Receptor 1 | |
100 μL | |
GPCR | |
134864 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |