missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAC-1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA5142114
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
TRAC-1 is a 232 amino acid protein containing an N-terminal RING-type zinc finger domain. It functions as a positive regulator in the T-cell receptor signaling pathway, thus leading to T-cell activation. It mediates ubiquitination and subsequent proteasomal degradation of target proteins. TRAC-1 is expressed in lymphoid tissues, including bone marrow, spleen, thymus as well as in other tissues. It is predominantly expressed in the CD4+ and CD8+ T-cells, suggesting that it is preferentially confined to T-cells.Especificaciones
TRAC-1 | |
Polyclonal | |
Unconjugated | |
RNF125 | |
4930553F04Rik; E3 ubiquitin-protein ligase RNF125; RING finger protein 125; ring finger protein 125, E3 ubiquitin protein ligase; RNF125; T-cell RING activation protein 1; T-cell ring protein identified in activation screen; TNORS; TRAC1; TRAC-1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
54941 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Flow Cytometry, Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96EQ8 | |
RNF125 | |
Peptide with sequence C-RSLLEYVNHSNTT. | |
100 μg | |
Primary | |
Human | |
Liquid |