missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAC-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA5106658
Detalles adicionales : Peso : 0.01000kg
Descripción
Antibody detects endogenous levels of total RNF125.
TRAC-1 is a 232 amino acid protein containing an N-terminal RING-type zinc finger domain. It functions as a positive regulator in the T-cell receptor signaling pathway, thus leading to T-cell activation. It mediates ubiquitination and subsequent proteasomal degradation of target proteins. TRAC-1 is expressed in lymphoid tissues, including bone marrow, spleen, thymus as well as in other tissues. It is predominantly expressed in the CD4+ and CD8+ T-cells, suggesting that it is preferentially confined to T-cells.Especificaciones
TRAC-1 | |
Polyclonal | |
Unconjugated | |
RNF125 | |
4930553F04Rik; E3 ubiquitin-protein ligase RNF125; RING finger protein 125; ring finger protein 125, E3 ubiquitin protein ligase; RNF125; T-cell RING activation protein 1; T-cell ring protein identified in activation screen; TNORS; TRAC1; TRAC-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
361296, 54941, 67664 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96EQ8, Q9D9R0 | |
RNF125 | |
A synthesized peptide derived from human RNF125(Accession Q96EQ8), corresponding to amino acid residues R138-L188. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |