missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAC-1 Monoclonal Antibody (1D3), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA522082
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide Sequence: FCQRELYEDS LLDHCITHHR SERRPVFCPL CRLIPDENPS SFSGSLIRHL QVSHTLFYDD FIDFNIIEEA LIRRVLDRSL LEYVNHSNT
TRAC-1 Monoclonal antibody specifically detects TRAC-1 in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Especificaciones
TRAC-1 | |
Monoclonal | |
0.34 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q96EQ8 | |
RNF125 | |
RNF125 (NP_060301, 143 a.a. ∽ 231 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunocytochemistry, Western Blot | |
1D3 | |
Unconjugated | |
RNF125 | |
4930553F04Rik; E3 ubiquitin-protein ligase RNF125; RING finger protein 125; ring finger protein 125, E3 ubiquitin protein ligase; RNF125; T-cell RING activation protein 1; T-cell ring protein identified in activation screen; TNORS; TRAC1; TRAC-1 | |
Mouse | |
Affinity chromatography | |
RUO | |
54941 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |