missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM85 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27708-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
May mediate anti-apoptotic activity.Especificaciones
TMEM85 | |
Polyclonal | |
Unconjugated | |
EMC4 | |
EMC4, ER membrane protein complex subunit 4, PIG17, TMEM85, transmembrane protein 85 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51234, 68032 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5J8M3, Q9CZX9 | |
EMC4 | |
TMEM85 Fusion Protein Ag26785 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |