Learn More
Tiomorfolina, con soporte polimérico, 0,8-1,1 mmol/g en resina de Merrifield, Thermo Scientific™
Catalog Number p-7047580
Click to view available options
Cantidad:
1 g
5 g
25 g
123-90-0
C4H9NS
103.183
MFCD03458715
BRNULMACUQOKMR-UHFFFAOYSA-N
thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine
67164
CHEBI:36392
tiomorfolina
C1CSCCN1
123-90-0
C4H9NS
103.183
MFCD03458715
BRNULMACUQOKMR-UHFFFAOYSA-N
thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine
67164
CHEBI:36392
tiomorfolina
C1CSCCN1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Thiomorpholine
Nombre de nota polymer-supported, 0.8 to 1.1mmol/g on Merrifield resin
Cantidad 1 g