Learn More
Nitrato de tetraetilamonio, solución acuosa al 35 % p/p, Thermo Scientific™
28.53€ - 107.00€
Identificadores de productos químicos
CAS | 1941-26-0 |
---|---|
Fórmula molecular | C8H20N2O3 |
Peso molecular (g/mol) | 192.26 |
Número MDL | MFCD00041978 |
Clave InChI | JTJKNAJRGLQKDZ-UHFFFAOYSA-N |
Sinónimo | tetraethylammonium nitrate, ethanaminium, n,n,n-triethyl-, nitrate, ammonium, tetraethyl-, nitrate, ethanaminium, n,n,n-triethyl-, nitrate 1:1, tetraethylazanium nitrate, tetraethyl ammonium nitrate, tetraethylammonium nitrate w/w aqueous solution |
PubChem CID | 74744 |
Nombre IUPAC | tetraethylazanium nitrate |
SMILES | [O-][N+]([O-])=O.CC[N+](CC)(CC)CC |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
1941-26-0 | |
192.26 | |
JTJKNAJRGLQKDZ-UHFFFAOYSA-N | |
74744 | |
[O-][N+]([O-])=O.CC[N+](CC)(CC)CC |
C8H20N2O3 | |
MFCD00041978 | |
tetraethylammonium nitrate, ethanaminium, n,n,n-triethyl-, nitrate, ammonium, tetraethyl-, nitrate, ethanaminium, n,n,n-triethyl-, nitrate 1:1, tetraethylazanium nitrate, tetraethyl ammonium nitrate, tetraethylammonium nitrate w/w aqueous solution | |
tetraethylazanium nitrate |
Especificaciones
Tetraethylammonium nitrate | |
1.029 g/mL | |
C8H20N2O3 | |
UN3139 | |
tetraethylammonium nitrate, ethanaminium, n,n,n-triethyl-, nitrate, ammonium, tetraethyl-, nitrate, ethanaminium, n,n,n-triethyl-, nitrate 1:1, tetraethylazanium nitrate, tetraethyl ammonium nitrate, tetraethylammonium nitrate w/w aqueous solution | |
[O-][N+]([O-])=O.CC[N+](CC)(CC)CC | |
192.26 | |
192.26 | |
Líquida |
1941-26-0 | |
>100°C | |
MFCD00041978 | |
3918466 | |
JTJKNAJRGLQKDZ-UHFFFAOYSA-N | |
tetraethylazanium nitrate | |
74744 | |
35% w/w aq. soln. |
Seguridad y manipulación
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H271-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING LIQUID, N.O.S.
missing translation for 'einecsNumber' : 217-725-5
missing translation for 'tsca' : Yes
Recommended Storage : Temperaturas ambientes