missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, ACS Reagent, ≥99%, Honeywell Riedel-de Haën™

Catalog Number p-7120866
Click to view available options
Cantidad:
1 L
2.5 L
Envase:
Glass Bottle
Este artículo no se puede devolver. Vea la política de devoluciones
127-18-4
C2Cl4
165.82
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
31373
CHEBI:17300
tetrachloroethene
ClC(Cl)=C(Cl)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
127-18-4
C2Cl4
165.82
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
31373
CHEBI:17300
tetrachloroethene
ClC(Cl)=C(Cl)Cl

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Tetrachloroethylene
Punto de fusión -22°C
Densidad 1.623g/mL at 25°C
Punto de ebullición 120°C to 122°C
Olor Characteristic
Envase Glass Bottle
Fórmula lineal CCl2=CCl2
Cantidad 1 L
Número UN UN1897
Beilstein 1361721
Densidad del vapor 5.83 (vs air)
Formula Weight (peso de la fórmula) 165.83g/mol
Presión de vapor 13mmHg (20°C), 19mmHg (25°C)
Porcentaje de pureza ≥99%
Forma física Liquid
Mostrar más Mostrar menos
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'