missing translation for 'onlineSavingsMsg'
Learn More

TER119 Rat, anti-Mouse, APC, Clone: TER-119, STEMCELL Technologies

Rat Monoclonal Antibody

Marca:  Stemcell Technologies 60033AZ

Detalles adicionales : Peso : 0.00110kg

 Ver más versiones de este producto

Código de producto. 17198651

  • 472.15€ / 100 microgramos
Fecha estimada de envío: 14-11-2022
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

  • The TER-119 antibody reacts with murine TER119 (Ly-76), an ∼52 kDa protein associated with glycophorin A on the surface of cells of the erythroid lineage in embryonic yolk sac, fetal and newborn liver, adult bone marrow, peripheral blood, and lymphoid organs
  • TER119 is an erythroid-specific marker expressed at all stages of differentiation from early proerythroblasts to mature erythrocytes, but not by erythroid colony-forming cells (BFU-E, blast-forming unit erythroid, or CFU-E, colony-forming unit erythroid)
  • The TER-119 antibody is a component of the 'lineage cocktail' used to detect, or deplete cells committed to hematopoietic lineages
  • In adult mice, TER119 is found on ∼20 - 25% of bone marrow cells and ∼2 - 3% of splenocytes
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD4+ T Cell Isolation Kit (Catalog #19852)
Especificaciones

Especificaciones

TER119
Monoclonal
APC
100 μg
Mouse
Erythroid Cells, Red Blood Cells
Immunofluorescence, Immunocytochemistry, Flow Cytometry, Western Blot, Immunohistochemistry
TER-119
Rat
Primary
IgG2b κ
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales