Learn More
Juchheim Laborgeraete™ Teclu Safety Burner
A high level of stability is achieved due to the high weight and low center of gravity of the burners.
Marca: Juchheim Laborgeraete™ 01374 DP
Detalles adicionales : Peso : 0.25000kg
Descripción
- For operation with option of natural gas or propane gas
- With needle valve
- With DVGW certification
- With air control screen
- Particularly good stand due to heavy, high-quality material
Especificaciones
Bunsen Burner | |
Brass | |
Needle Valve, Safety Valve, and Automatic Gas shut-off |
Propane | |
Gas consumption approx. 39 L/h (0.0787 kg/h), Heating capacity approx. 3810 kJ/h |