missing translation for 'onlineSavingsMsg'
Learn More

TEA, solución de tampón de 0,2 M, pH 7,0, Thermo Scientific™

Catalog Number 15424139
Click to view available options
Cantidad:
250 mL
500 mL
Este artículo no se puede devolver. Vea la política de devoluciones
102-71-6
C6H15NO3
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
Triethanolamine buffer solution
7618
CHEBI:28621
2-[bis(2-hidroxietil)amino]etanol
C(CO)N(CCO)CCO
Este artículo no se puede devolver. Vea la política de devoluciones
102-71-6
C6H15NO3
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
Triethanolamine buffer solution
7618
CHEBI:28621
2-[bis(2-hidroxietil)amino]etanol
C(CO)N(CCO)CCO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material TEA buffer soln.
Concentración 0.2 M
pH 7.0
Forma física Líquida
Cantidad 500 mL
Olor Odorless
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'