missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAOK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA124577
Detalles adicionales : Peso : 0.01000kg
Description
PA1-24577 detects Serine/threonine kinase TAO kinase 1 in Human samples.
TAOK1 is a serine/threonine-protein kinase involved in regulation of the p38-containing stress-responsive MAP kinase pathway and extracellular signal-regulated protein kinase (ERK) kinases (MEKs). The activation of and binding to MEK3 by TAOK1 implicates TAOK1 in the regulation of the p38-containing stress-responsive MAP kinase pathway. A microtubule affinity-regulating kinase, TAOK1 (also known as MARKK) is an important regulator of mitotic progression, required for both chromosome congression and checkpoint-induced anaphase delay. TAOK1 is involved in various processes such as p38/MAPK14 stress-activated MAPK cascase, DNA damage response, and regulations of cytoskeleton stability. It phosphorylates MAP2K3, MAP2K6, and MARK2. TAOK1 acts as an activator of the p38/MAPK cascade by mediating phosphorylation and subsequent activation of the upsteam MAP2K3 and MAP2K6 kinases. It is involved in G-protein couples receptor signaling to p38/MAPK14. In response to DNA damage, TAOK1, is involved in the G2/M transition DNA damage checkpoint by activating the p38/MAPK14 stress-activated MAPK cascade by phosphorylaton of MAP2K3 and MAP2K6. In addition, it acts as a regulator of cytoskeleton stability and a regulator of apoptosis.Specifications
TAOK1 | |
Polyclonal | |
Unconjugated | |
TAOK1 | |
2810468K05Rik; AU020252; D130018F14Rik; FLJ14314; h hTAOK1; H KIAA1361; hKFC-B; hTAOK1; KFC-B; Kiaa1361; Kinase from chicken homolog B; MAP3K16; MARK Kinase; Markk; microtubule affinity regulating kinase kinase; mKIAA1361; prostate-derived STE20-like kinase 2; prostate-derived sterile 20-like kinase 2; PSK2; PSK-2; serine/threonine protein kinase TAO1; serine/threonine protein kinase TAO1 homolog; serine/threonine-protein kinase TAO1; STE20-like kinase PSK2; tao; TAO kinase 1; TAO1; TAOK1; Thousand and one amino acid protein 1; thousand and one amino acid protein kinase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57551 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q7L7X3 | |
TAOK1 | |
Synthetic 16 amino acid peptide from internal region of human TAOK1 | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |