missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAAR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA132858
Detalles adicionales : Peso : 0.01000kg
Descripción
PA1-32858 detects GPCR GPR58 from human samples.
Olfactory receptor activated by several primary trace amines, including isoamylamine. Activated by isoamylamine and cyclohexylamine, but not to the corresponding alcohols, isoamylalcohol and cyclohexanol. This receptor is probably mediated by the G(s)-class of G-proteins which activate adenylate cyclase. [GeneCards]Especificaciones
TAAR3 | |
Polyclonal | |
Unconjugated | |
TAAR3P | |
GPR57; GPR58; mTaar3; TAAR2; Taar3; TaR-3; trace amine associated receptor 3; trace amine associated receptor 3 (gene/pseudogene); trace amine receptor 3; trace amine-associated receptor 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9288 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9P1P4 | |
TAAR3P | |
Synthetic 18 amino acid peptide from C-terminus of human GPCR GPR58 | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |