missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAU2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15998-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
STAU2 (Staufen homolog 2) is a member of the family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. Staufen homolog 2 shares 48.5% and 59.9% similarity with drosophila and human staufen, respectively. The exact function of Staufen homolog 2 is not known, but since it contains 3 copies of conserved dsRNA binding domain, it could be involved in double-stranded RNA binding events. Several transcript variants encoding different isoforms have been found for this gene.Especificaciones
STAU2 | |
Polyclonal | |
Unconjugated | |
STAU2 | |
39K2, 39K3, STAU2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171500, 27067, 29819 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68SB1, Q8CJ67, Q9NUL3 | |
STAU2 | |
STAU2 Fusion Protein Ag8482 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |