missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StAR (Steroidogenic Acute Regulator) (Leydig Cell Marker) Mouse, Clone: STAR/2140, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 6770-MSM3-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
StAR (Steroidogenic Acute Regulator) (Leydig Cell Marker) | |
Cytoplasmic (Mitochondria) | |
STAR/2140 | |
Unconjugated | |
Mouse | |
30kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
6770 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment (apprx. aa 39-108) of human STAR protein | |
K-562 cells. Adrenal or Testicular Carcinoma | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |