Learn More
De Buyer™ Recipiente de pastelería laboratorio de acero inoxidable
Marca: De Buyer™ 325026
Detalles adicionales : Peso : 0.41000kg
Especificaciones
Pastry Bowl | |
For making mixtures, mixing, Whisking, heating by bain-marie, for storing a preparation or ingredients | |
120 mm | |
Flat bottom | |
4.5 L |
Stainless Steel | |
Fridge, deep-freezer, blast-chiller and oven | |
Each | |
Sterile |