Learn More
Gasa de alambre de acero inoxidable, malla 20 tejida a partir de alambre de 0,382 mm de diámetro, tipo 304, Thermo Scientific Chemicals
Filtración, electrodos
Marca: Thermo Scientific Alfa Aesar 040942.FL
Detalles adicionales : Peso : 0.01000kg
Descripción
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Stainless Steel gauze, Type 304 | |
Fe:Cr:Ni; 70:19:11 wt% | |
75 x 75 mm | |
20 mesh |
Woven from 0.382mm (0.015 in.) dia. wire | |
MFCD00148499 | |
Insoluble in water. | |
Tela de alambre |
RUO – Research Use Only