Learn More
Buerkle™ Stackable Graduated Beakers
Click to view available options
Capacidad (métrico):
3000 mL
Descripción
- Graduation printed in blue
- Open handle
- Conical
- Transparent polypropylene
- Sterilizable
- Shatterproof
- Good chemical resistance
Especificaciones
Color | Transparent |
Capacidad (métrico) | 3000 mL |
Material | Polypropylene |
Graduated | Yes |
Modelo | Conical |