Learn More
Buerkle™ Stackable Graduated Beakers
Catalog Number 11822293
Click to view available options
Capacidad (métrico):
3000 mL
These beakers can be stacked and therefore save space during storage.
  • Graduation printed in blue
  • Open handle
  • Conical
  • Transparent polypropylene
  • Sterilizable
  • Shatterproof
  • Good chemical resistance
Color Transparent
Capacidad (métrico) 3000 mL
Material Polypropylene
Graduated Yes
Modelo Conical