missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate Solution, 1M, Honeywell Fluka™
missing translation for 'orderingAttributeHoverText'
Cantidad:
1 L
20 L
6 x 1 L
Beschreibung
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Spezifikation
Nombre del producto químico o material | Sodium thiosulfate solution |
Cantidad | 20 L |
Fórmula lineal | Na2S2O3 |
Número UN | NONH for all modes of transport |
Nombre IUPAC | disodium sulfanidesulfonate |
Formula Weight (peso de la fórmula) | 158.11g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'