missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate Solution, 0.0394M, Honeywell Fluka™
Click to view available options
Cantidad:
1 L
5 L
10 L
6 x 1 L
Descripción
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Especificaciones
Nombre del producto químico o material | Sodium thiosulfate solution |
Cantidad | 10 L |
Fórmula lineal | Na2S2O3 |
Número UN | NONH for all modes of transport |
Nombre IUPAC | disodium sulfanidesulfonate |
Formula Weight (peso de la fórmula) | 158.11g/mol |
Densidad | 1.01g/mL at 20°C |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'