missing translation for 'onlineSavingsMsg'
Learn More

Sulfato de sodio, ACS, 99,0 % mín., Thermo Scientific Chemicals

Catalog Number p-4407937
Click to view available options
Cantidad:
500 g
2 kg
10 kg
Este artículo no se puede devolver. Vea la política de devoluciones
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
disodio; sulfato
[Na+].[Na+].[O-]S([O-])(=O)=O
Este artículo no se puede devolver. Vea la política de devoluciones
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
disodio; sulfato
[Na+].[Na+].[O-]S([O-])(=O)=O

Para su uso en el secado de líquidos orgánicos, en la determinación de nitrógeno Kjeldahl, en la fabricación de vidrio

In drying organic liquids, in Kjeldahl nitrogen determination, in manufacturing of glass. Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Sodium sulfate
Punto de fusión 884°C
Forma física Polvo
Cantidad 500 g
Índice Merck 14,8680
Información de solubilidad Soluble in water; Insoluble in alcoholSoluble in water,glycerol and hydrogen iodide; Insoluble in ethanol and many organic solvents.
Formula Weight (peso de la fórmula) 142.04
Porcentaje de pureza ≥99.0%
Grado ACS
Olor Odorless
Índice de refracción 1.484
Sensibilidad Hygroscopic
Densidad 2.680
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'