missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10379-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein belongs to the small nuclear ribonucleoprotein core protein family. It is required for pre-mRNA splicing and small nuclear ribonucleoprotein biogenesis.Especificaciones
SNRPD3 | |
Polyclonal | |
Unconjugated | |
SNRPD3 | |
Sm D3, SMD3, snRNP core protein D3, SNRPD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6634, 67332, 687711 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P62318, P62320 | |
SNRPD3 | |
SNRPD3 Fusion Protein Ag0293 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |