missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14465-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Members of the parvin family, including PARVA, are actin-binding proteins associated with focal contacts.Especificaciones
SMC6 | |
Polyclonal | |
Unconjugated | |
SMC6 | |
hSMC6, SMC 6, SMC protein 6, SMC6, SMC6L1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313961, 67241, 79677 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q924W5, Q96SB8 | |
Smc6 | |
SMC6 Fusion Protein Ag5812 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |