missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC44A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14687-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
SLC44A1 encodes a choline transporter and may be involved in membrane synthesis and myelin production.Especificaciones
SLC44A1 | |
Polyclonal | |
Unconjugated | |
SLC44A1 | |
CD92, CDW92, choline transporter-like protein 1, CHTL1, CTL1, SLC44A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100434, 23446, 85254 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6X893, Q8VII6, Q8WWI5 | |
Slc44a1 | |
SLC44A1 Fusion Protein Ag6024 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |