missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC20A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12820-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the inorganic phosphate transporter family. The encoded protein is a type 3 sodium-dependent phosphate symporter that plays an important role in phosphate homeostasis by mediating cellular phosphate uptake. The encoded protein also confers susceptibility to viral infection as a gamma-retroviral receptor. Mutations in this gene may play a role in familial idiopathic basal ganglia calcification. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Especificaciones
SLC20A2 | |
Polyclonal | |
Unconjugated | |
SLC20A2 | |
Glvr 2, GLVR2, hPit2, MLVAR, Phosphate transporter 2, PIT 2, Pit2, PiT-2, SLC20A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20516, 29502, 6575 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q08357, Q63488, Q80UP8 | |
SLC20A2 | |
SLC20A2 Fusion Protein Ag3469 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |