missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Sulfate, ACS Reagent, Honeywell Fluka™
Click to view available options
Cantidad:
10 g
100 g
500 g
Descripción
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Especificaciones
Nombre del producto químico o material | Silver Sulfate |
Cantidad | 100 g |
Fórmula lineal | Ag2SO4 |
Número UN | 3077 |
Formula Weight (peso de la fórmula) | 311.8g/mol |
Porcentaje de pureza | 99% |
Grado | ACS Reagent |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'