missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluoroantimonato de plata, 99 %, Thermo Scientific Chemicals
Click to view available options
Cantidad:
1 g
5 g
25 g
Descripción
Silver hexafluoroantimonate is used as a catalyst for the preparation of a number of compounds. It acts as an acidic catalyst in epoxide opening reactions and sulfenylation reactions , controlled initiator in polymerization reactions, and as an activator of catalyst in certain organic reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Silver hexafluoroantimonate |
Color | Blanco a crema |
Forma física | Polvo/masa |
Cantidad | 5 g |
Número UN | UN3260 |
Información de solubilidad | Soluble in water. |
Formula Weight (peso de la fórmula) | 343.41 |
Porcentaje de pureza | 99% |
Sensibilidad | Hygroscopic; Light sensitive |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'