missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGIRR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27828-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
SIGIRR (single Ig IL-1R-related molecule) is a novel member of the interleukin 1 receptor (IL-1R) superfamily. Although it shows the typical conserved motifs that characterize the IL-1R and Toll superfamily, it is structurally and functionally distinct from both. SIGIRR has only one Ig domain in its extracellular portion whereas the IL-1R family contains three Ig folds. An unusually long cytoplasmic domain is reminiscent of the structure of drosophila Toll, yet the SIGIRR peptide sequence is more closely related to IL-1RI. A chimera, in which the SIGIRR intracellular domain was fused to the IL-1R extracellular domain, did not activate NFkappaB unlike similar fusion proteins of other IL-1R related molecules. Phylogenetic data that suggest that the plasma membrane protein SIGIRR is a close functional relative of MyD88 in mammals.Especificaciones
SIGIRR | |
Polyclonal | |
Unconjugated | |
SIGIRR | |
SIGIRR, TIR8, Toll/interleukin 1 receptor 8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24058, 59307 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6IA17, Q9JLZ8 | |
SIGIRR | |
SIGIRR Fusion Protein Ag25850 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |