Learn More
SEC31B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA557445
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: HAQGSAVLGQ QSPPFPFPRI VVGATLHSKE TSSYRLGSQP SHQVPTPSPR PRVFTPQSSP AMPLAPSHPS PYQGPRTQNI SDYRA Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 61%.
This gene encodes a protein of unknown function. The protein has moderate similarity to rat VAP1 protein which is an endosomal membrane-associated protein, containing a putative Ca2+/calmodulin-dependent kinase II phosphorylation site.Especificaciones
SEC31B | |
Polyclonal | |
Unconjugated | |
SEC31B | |
Protein transport protein Sec31B; sc31b; SEC31 homolog B; SEC31 homolog B, COPII coat complex component; SEC31 homolog B, COPII coating complex component; SEC31B; SEC31B-1; SEC31L2; SEC31-like 2; SEC31-like protein 2; SEC31-related protein B; secretory pathway component Sec31B-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25956 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NQW1 | |
SEC31B | |
Recombinant protein corresponding to Human SEC31B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only