Learn More
SEC24A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558582
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: FLPEANLPPP LNWQYNYPST ASQTNHCPRA SSQPTVSGNT SLTTNHQYVS SGYPSLQNSF IKSGPSVPPL VNPPLPTTFQ PGA Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 64%.
SC24A is a component of coat protein II (COPII; see MIM 601924)-coated vesicles that mediate protein transport from the endoplasmic reticulum. SEC24A is 1 of several mammalian proteins that show structural and functional homology to yeast Sec24.Especificaciones
SEC24A | |
Polyclonal | |
Unconjugated | |
SEC24A | |
Protein transport protein Sec24A; SEC24 family member A; SEC24 family, member A; SEC24 homolog A, COPII coat complex component; SEC24 related gene family, member A; Sec24 related gene family, member A (S. cerevisiae); Sec24a; SEC24-related protein A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10802 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95486 | |
SEC24A | |
Recombinant protein corresponding to Human SEC24A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only