Learn More
SDHB Mouse, anti-Human, CoraLite 488, Clone: 1A2A5, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech CL488-67600-100UL
Detalles adicionales : Peso : 0.00020kg
Código de producto. 17231566
  • 367.00€ / 100 microlitros
Fecha estimada de envío: 14-05-2024
para ver el stock
Descripción
Descripción
SDHB(Succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial) is also named as SDH, SDH1 and belongs to the succinate dehydrogenase/fumarate reductase iron-sulfur protein family. The SDHD, SDHB, and SDHC genes encode subunits of mitochondrial complex II (succinate dehydrogenase). Mitochondrial complex II is a heterotetrameric complex involved in the aerobic electron transport chain and catalyses the oxidation of succinate to fumarate (Krebs cycle) with transport of electrons to the ubiquinone pool. SDHA and SDHB are the hydrophilic catalytic part of the complex and are highly conserved(J Med Genet 2004;41:e99). Defects in SDHB are a cause of susceptibility to pheochromocytoma (PCC), paragangliomas type 4 (PGL4), paraganglioma and gastric stromal sarcoma (PGGSS) and Cowden-like syndrome (CWDLS).
Especificaciones
Especificaciones
SDHB
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat
Immunofluorescence
1A2A5
CoraLite 488
100 μL
6390