Learn More
Thermo Scientific Chemicals Sustrato de zafiro 10 x 10 x 0,432 mm, EPI pulido por dos lados, eje C.
Marca: Thermo Scientific Chemicals 045017.KT
Detalles adicionales : Peso : 0.01000kg
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Sustrato de zafiro | |
EPI pulido por dos lados | |
1 unidad | |
10 x 10 x 0,432mm |
RUO – Research Use Only