missing translation for 'onlineSavingsMsg'
Learn More

Diclorhidrato de ácido (S)-(-)-piperazina-2-carboxílico, + 98 %, Thermo Scientific Chemicals

Catalog Number 15477745
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
158663-69-5
C5H12Cl2N2O2
203.06
MFCD01632104,MFCD03840829
WNSDZBQLMGKPQS-UHFFFAOYNA-N
s-piperazine-2-carboxylic acid dihydrochloride, s-2-piperazinecarboxylic acid dihydrochloride, 2s-piperazine-2-carboxylic acid dihydrochloride, s-piperazine-2-carboxylic acid 2hcl, s---2-piperazinecarboxylic acid dihydrochloride, s---piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, 2s, s---piperazine-2-carboxylic acid 2hcl, 2s---2-carboxypiperazine dihydrochloride
2734260
ácido (2S)-piperazina-2-carboxílico; diclorhidrato
Cl.Cl.OC(=O)C1CNCCN1
Este artículo no se puede devolver. Vea la política de devoluciones
158663-69-5
C5H12Cl2N2O2
203.06
MFCD01632104,MFCD03840829
WNSDZBQLMGKPQS-UHFFFAOYNA-N
s-piperazine-2-carboxylic acid dihydrochloride, s-2-piperazinecarboxylic acid dihydrochloride, 2s-piperazine-2-carboxylic acid dihydrochloride, s-piperazine-2-carboxylic acid 2hcl, s---2-piperazinecarboxylic acid dihydrochloride, s---piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, 2s, s---piperazine-2-carboxylic acid 2hcl, 2s---2-carboxypiperazine dihydrochloride
2734260
ácido (2S)-piperazina-2-carboxílico; diclorhidrato
Cl.Cl.OC(=O)C1CNCCN1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material (S)-(-)-Piperazine-2-carboxylic acid dihydrochloride
Punto de fusión ∽280°C (decomposition)
Cantidad 250 mg
Sensibilidad Hygroscopic
Rotación óptica −5° (c=1 in Water)
Formula Weight (peso de la fórmula) 203.07
Porcentaje de pureza ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'