Learn More
RXFP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA589738
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: CGDTSGWATI FGTVHGNANS VALTQECFLK QYPQCCDCKE TELECVNGDL KSVPMISNNV TLLSLKKNKI HSLPDKVFIK YTKLKKIFLQ HNCIRHISRK A; Positive Samples: Mouse kidney, Rat kidney; Cellular Location: Cell membrane, Multi-pass membrane protein
The activity of this relaxin receptor is mediated by G proteins leading to stimulation of adenylate cyclase and an increase of cAMP and may also be a receptor for Leydig insulin-like peptide. Expressed in embryonic and adult gonads of males and females, as well in male gubernarculum and in brain. Not detected in kidney, spleen and heart.Especificaciones
RXFP2 | |
Polyclonal | |
Unconjugated | |
RXFP2 | |
G protein coupled receptor affecting testicular descent, G protein-coupled receptor 106, Gpcr, GPR106, G-protein coupled receptor 106, G-protein coupled receptor affecting testicular descent, GREAT, INSL3 receptor, INSL3R, leucine-rich repeat-containing G protein-coupled receptor 8, leucine-rich repeat-containing G-protein coupled receptor 8, Lgr8, LGR8.1, Relaxin family peptide receptor 2, relaxin receptor 2, relaxin/insulin like family peptide receptor 2, relaxin/insulin-like family peptide receptor 2, Rxfp2, RXFPR2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
140498, 363866 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.44 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
Q91ZZ5 | |
RXFP2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 80-180 of human RXFP2 (NP_5707181) | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |