missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXFP2 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA577719
Detalles adicionales : Peso : 0.01000kg
Descripción
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
The activity of this relaxin receptor is mediated by G proteins leading to stimulation of adenylate cyclase and an increase of cAMP and may also be a receptor for Leydig insulin-like peptide. Expressed in embryonic and adult gonads of males and females, as well in male gubernarculum and in brain. Not detected in kidney, spleen and heart.Especificaciones
RXFP2 (extracellular) | |
Polyclonal | |
Unconjugated | |
Rxfp2 | |
Rxfp2, G-protein coupled receptor 106, G-protein coupled receptor affecting testicular descent, Leucine-rich repeat-containing G-protein coupled receptor 8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140498, 363866 | |
-20°C | |
Lyophilized |
Western Blot | |
0.85 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q91ZZ5 | |
Rxfp2 | |
Peptide CFPLHYDQAEDFGSR, corresponding to amino acid residues 556-570 of mouse Relaxin Receptor 2 | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |