Learn More
RUFY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19180-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
RUFY2 (RUN And FYVE Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is RUNDC3B.Especificaciones
RUFY2 | |
Polyclonal | |
Unconjugated | |
RUFY2 | |
KIAA1537, RABIP4R, RUFY2, ZFYVE13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55680, 690777, 70432 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R4C2, Q8WXA3 | |
RUFY2 | |
RUFY2 Fusion Protein Ag5938 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |