missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSPH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17603-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Especificaciones
RSPH3 | |
Polyclonal | |
Unconjugated | |
RSPH3 | |
RSHL2, RSP3, RSPH3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361476, 83861 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86UC2 | |
Rsph3 | |
RSPH3 Fusion Protein Ag11707 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |