Learn More
RNF14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA529456
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Recommended positive controls: IMR32, U87-MG. Predicted reactivity: Mouse (92%), Rat (90%), Rhesus Monkey (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein interacts with androgen receptor (AR) and may function as a coactivator that induces AR target gene expression in prostate. A dominant negative mutant of this gene has been demonstrated to inhibit the AR-mediated growth of prostate cancer. This protein also interacts with class III ubiquitin-conjugating enzymes (E2s) and may act as a ubiquitin-ligase (E3) in the ubiquitination of certain nuclear proteins. Five alternatively spliced transcript variants encoding two distinct isoforms have been reported.Specifica
RNF14 | |
Polyclonal | |
Unconjugated | |
RNF14 | |
2310075C09Rik; 2610005D23Rik; AA986456; androgen receptor associated protein 54; androgen receptor associated protein 54 (androgen receptor coactivator); androgen receptor-associated protein 54; ARA54; AU041447; D18Ertd188e; D7Bwg0165e; E3 ubiquitin-protein ligase RNF14; HFB30; HRIHFB2038; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase RNF14; protein Triad2; RING finger protein 14; RNF14; TRIAD2; Triad2 protein; TRIAD2 type I | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9604 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9UBS8 | |
RNF14 | |
Recombinant fragment corresponding to a region within amino acids 1 and 260 of Human RNF14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |