Learn More
Fisherbrand™ Remote Dispenser with Sensor
Includes a built-in high precision resistivity sensor with cell constant of 0.01 cm-1. Fisher Scientific™ Remote Dispenser with Sensor can measure the quality of ultrapure water just prior to the dispensing point.
Marca: Fisherbrand™
Detalles adicionales : Peso : 1.00000kg
Descripción
- Point of Use remote water dispenser
- Built-in high precision resistivity sensor with cell constant of 0.01cm-1 to measure the quality of ultrapure water just prior to the dispensing point
- Volumetric dispensing and calibration
- 360 degree-rotatable and detachable handle
- Effortless and instant dispensing with maximum flexibility
Especificaciones
Accu500 | |
Remote Water Dispenser |