missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Rhesus Macaque IL-5 Protein

Código de producto. 18667676 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
500 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
500 microgramos
Product Code. Cantidad unitSize
18667676 10 μg 10 microgramos
18640038 100 μg 100 microgramos
18682187 1 mg 1 miligramo
18611347 500 μg 500 microgramos
4 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 18667676

missing translation for 'mfr': Novus Biologicals™ NBP24138710ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

IL-5 protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 4 ng/mL, corresponding to a specific activity of > 2.5 x 10^5 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Número de acceso P48093, P48093, P48093
Para utilizar con (aplicación) SDS-PAGE, Bioactivity
Formulación Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4.
ID de gen (Entrez) 3567
Peso molecular MolecularWeight-theroretical: 26.1 kDa
Cantidad 10 μg
Requisitos de almacenamiento Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Concentración de endotoxinas Less than 0.1 EU/μg of IL-5 AS determined by LAL method.
Alias de gen B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5 T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I
Símbolo de gen IL5
Reconstitución Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Categoría de investigación Biologically Active Proteins
Grado de pureza o calidad >98 % pure by SDS-PAGE and HPLC
Expresión IL-5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.