missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CCL22 / MDC Protein
A DNA sequence encoding the Rat CCL22 / MDC, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10298-ec-20ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
CCL22 / MDC Protein | |
Research Use Only | |
CCL22 | |
The biologically active determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration range of 10-100 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GPYGANVEDS ICCQDYIRHP LPPRFVKEFY WTSKSCRKPG VVLITIKNRD ICADPRMLWV KKILHKLA | |
Greater than 96% as determined by SDS-PAGE and HPLC. |