missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Protein A His (C-Term) Cys (C-Term) Protein

Código de producto. p-200054606 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
10 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
Código de producto. Cantidad unitSize
18486811 100 mg 100 miligramos
18452222 10 mg 10 miligramos
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18486811

Marca: Novus Biologicals™ NBP235014100mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

Protein A is a genetically engineered protein containing 5 IgG-binding regions of protein A. Cell wall binding region, cell membrane binding region and albumin binding region have been removed from the recombinant Protein A to ensure the maximum specific IgG binding. The recombinant Protein A is ideal for purification of polyclonal or monoclonal IgG antibodies. Protein A binds to various human, mouse and rat IgG subclasses (e.g., human IgG1, IgG2, IgG4; mouse IgG2a, IgG2b, IgG3; rat IgG2c). It also binds to
TRUSTED_SUSTAINABILITY

Specifications

Concentración LYOPH
Para utilizar con (aplicación) PAGE, Bioactivity
Formulación Lyophilized from additive free solution.
ID de gen (Entrez) 3919448
Peso molecular 34.8 kDa
Cantidad 100 mg
Requisitos de almacenamiento Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Concentración de endotoxinas Less than 0.1 EU/ug of Protein A as determined by LAL method.
Alias de gen Immunoglobulin G binding protein A, SPA, Staphylococcal protein A
Reconstitución Reconstitute with distilled water or saline.
Categoría de investigación Biologically Active Proteins, Epitope Tags
Grado de pureza o calidad >95% by SDS-PAGE and HPLC
Expresión Protein A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.