Learn More
Novus Biologicals™ Recombinant PreScission Protease Protein
Código de producto. 18602248 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 UI
250 UI
Tamaño de la unidad:
cada uno
18602248 250 UI cada uno
18649418 100 UI cada uno
2 options
Código de producto. 18602248
Marca: Novus Biologicals™ NBP261442250IU
Recombinant Protein
The protease specifically recognizes a subset of sequences which include the core amino acid sequence Leu-Phe-Gln/Gly-Pro cleaving between the Gln and Gly residues. Substrate recognition and cleavage are likely to be dependent not only upon primary structural signals, but also upon the super structures of the fusion protein.
TRUSTED_SUSTAINABILITY
Especificaciones
Para utilizar con (aplicación) SDS-PAGE, Enzyme Activity
Formulación 50 mM Tris-HCl, pH 7.0, 150 mM NaCl, 1 mM EDTA, and 1 mM DTT.
Peso molecular MolecularWeight-theroretical: 43 kDa
Cantidad 250 UI
Requisitos de almacenamiento Store at -20°C. Avoid freeze/thaw cycles.
Alias de gen 3 C protease, Picornain 3 C, PSP
Propiedades del inhibidor During cleavage reactions, it is recommended that samples be removed at various time points and analyzed by SDS-PAGE to estimate the yield, purity, and extent of digestion. The amount of PreScission Protease, temperature and length of incubation required for complete digestion of a given GST fusion partner may vary depending on the fusion partner. It works most effective at 4 C but can digest substrates (at room temperature. Optimal conditions for each fusion should be determined in pilot experiments. Digestion may be improved by adding TritonTM X-100, TweenTM 20 or NonidetTM P40 to a concentration of 0.01 %. Concentrations of these detergents up to 1 % do not inhibit PreScission Protease.)
Grado de pureza o calidad Peptide affinity purified
Expresión PreScission Protease