missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Antibacterial peptide PMAP-36 Protein
A DNA sequence encoding the Sus scrofa (Pig) Antibacterial peptide PMAP-36, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7219-ec-50ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001123437.1 | |
Antibacterial peptide PMAP-36 Protein | |
Research Use Only | |
PMAP36 | |
Recombinant Protein | |
E. coli | |
VGRFRRLRKK TRKRLKKIGK VLKWIPPIVG SIPLGCG | |
Greater than 90% as determined by SDS-PAGE. |
100170124 | |
50 μg | |
Not determined. | |
Full Length | |
Pig | |
His-SUMO | |
Tris-based buffer, 50% glycerol |