Learn More
enQuireBio™ Recombinant other MT I Protein
Recombinant Protein
Marca: enQuireBio™ QP12745-100mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MT I Protein | |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |
Greater than 98.0% as determined by RP-HPLC. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein (1 mg/ml) was lyophilized with no additives. |